Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ergodyne™ ProFlex™ 901 Half-Finger Leather Impact Gloves

Catalog No. 19820237 Shop All Ergodyne Products
Change view
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19-820-237 Large
19-820-235 Small
19-820-236 Medium
19-820-239 2X-Large
19-820-238 X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Catalog No. 19-820-237 Supplier Ergodyne™ Supplier No. 17784
Add to Cart
Add to Cart
This item is not returnable. View return policy

Foam padding in the palm and other key areas helps to reduce shock and impact. genuine leather construction provides a natural, yet durable feel.

  • Segmented foam palm padding reduces shock and impact
  • Genuine leather palm provides durability with a natural feel
  • Half-finger design for dexterity and tactile feedback
  • Hex stretch knit and breathable spandex construction
  • Finger webbing loops for easy on/off
  • Neoprene knuckle pad
  • Low-profile molded hook & loop closure
  • Gloves feature ID space to write user's name

Specifications

Back Material Spandex
Color Black
Hand Specific
Material Geniune Leather
Product Type Anti-impact Gloves
Size Large
For Use With (Application) Construction, Manufacturing, Hand and Power Tools
Glove Cuff Style Hook and Loop
Series ProFlex™ 901
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.