Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ergodyne™ ProFlex™ 7141 Hi-Vis Nitrile-Coated Cut-Resistant Gloves

Catalog No. 19820262 Shop All Ergodyne Products
Encompass_Preferred
Change view
Click to view available options
Size:
Small
Medium
Large
2X-Large
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19-820-262 2X-Large
19-820-258 Small
19-820-259 Medium
19-820-260 Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Catalog No. 19-820-262 Supplier Ergodyne™ Supplier No. 17916
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Designed with two new technologies to create comfortable, breathable and secure grip ANSI cut level 4 gloves. Features molded TPR Armor™ on the knuckles and fingers for dorsal impact protection.

  • Made with high-performance polyethylene (HPPE) fibers for A4 cut protection
  • HPPE does not include steel or glass often used in other anti-cut gloves which can cause irritation
  • WSX™ dip combined with TenaLux™ yarn make these palm-coated gloves cool to the touch with 360-degrees of breathability
  • Lighter weight than traditional A4 cut-resistant gloves
  • Hi-vis lime color on the backside of the hand
  • Liquid-resistant special coating absorbs and wicks moisture from the surface allowing for a firm-grip in wet oily and wet conditions
  • Water-based polyurethane (PU) coating combined with nitrile dip allows maximum dexterity and significantly relieves hand fatigue while ensuring durability
  • Molded TPR Armor™ on the backside of the glove provides pinch injury prevention and blow defection
  • TPR™ extends to fingertips and is sewn on for maximum durability
  • WearArmor™ flex zones deliver protection without sacrificing dexterity or flexibility
  • Ideal for workers in heavy construction, oil and gas, mining, demolition, extraction and refining, ironwork or cargo handling

Specifications

ANSI Abrasion Rating ANSI 4
Construction Seamless Knit
Coating Material Nitrile
Color Lime Green, Black
Cut Resistance Rating ANSI A4
Hand Specific
Material HPPE
Product Type Cut-resistant Gloves
Size 2X-Large
For Use With (Application) Construction, assembly, fabrication, pulp and paper, service technicians
Grip Design WSX dip technology
Model 17916
Series ProFlex™ 7141
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.