Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ergodyne™ ProFlex™ 200 Short Light Duty Copolymer Knee Pads

Catalog No. 19820273 Shop All Ergodyne Products
Encompass_Preferred
Change view
Click to view available options
copolymer knee pads
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
19-820-273 copolymer knee pads
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 19-820-273 Supplier Ergodyne™ Supplier No. 18200
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Economy lightweight comfort. Single strap.

Specifications

Color Black
For Use With (Application) Construction, Maintenance, Flooring,Landscaping, Mechanics, Trades
Liner Material Copolymer
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.