Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ergodyne™ ProFlex™ 4000 Single Strap Wrist Support

Catalog No. 19910431 Shop All Ergodyne Products
Encompass_Preferred
Change view
Click to view available options
Color:
Black
Tan
Size:
Small
Large
X-Large
Hand:
Left
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color Size Hand
19-910-431 Black X-Large Left
19-910-430 Black Small Left
19-910-437 Tan Large Left
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 19-910-431 Supplier Ergodyne™ Supplier No. 70018
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Single strap support with open center supports the wrist during repetitive motion discomfort by limiting wrist movements.

Specifications

Color Black
Size X-Large
For Use With (Application) Food Processing,Picking/Packing,Assembly/Fabrication
Hand Left
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.