Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ergodyne™ ProFlex™ 340 Slip Resistant Knee Pads with Shin Guard

Catalog No. 19820280 Shop All Ergodyne Products
Encompass_Preferred
Change view
Click to view available options
Material:
840D Nylon
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Material
19-820-280 840D Nylon
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 19-820-280 Supplier Ergodyne™ Supplier No. 18340
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Grips wet or dry work surfaces and offers extra protection for shins.

Specifications

Color Black Cap
For Use With (Application) Construction, Masonry, Freight/Baggage,Flooring, Maintenance, Utilities, Carpentry
Material 840D Nylon
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.