Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ergodyne™ ProFlex™ 821 Smooth Surface Handling Work Gloves

Catalog No. 19820118 Shop All Ergodyne Products
Encompass_Preferred
Change view
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19-820-118 Large
19-820-116 Small
19-820-117 Medium
19-820-119 X-Large
19-820-120 2X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Catalog No. 19-820-118 Supplier Ergodyne™ Supplier No. 17234
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Palm and index fingers feature a silicone coating to provide a secure grip when handling smooth surfaces such as boxes or cartons.

  • Breathable polyester mesh keeps hands cool without being bulky or stiff
  • Low-profile, molded hook & loop closure provides a secure fit
  • Gloves feature an ID tab for personalization to prevent misplaced gloves
  • Pull-on tab for easy on/off
  • Machine washable

Specifications

Coating Material Silicone
Color Black
Hand Specific
Material Polyester Mesh
Product Type Work Gloves
Size Large
For Use With (Application) Warehousing/Distribution, Package Delivery
Glove Cuff Style Open
Series ProFlex™ 821
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.