Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ergodyne™ ProFlex™ 4020 Wrist Support

Catalog No. 19910447 Shop All Ergodyne Products
Encompass_Preferred
Change view
Click to view available options
Hand:
Left
Right
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Hand
19-910-447 Left
19-910-446 Right
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 19-910-447 Supplier Ergodyne™ Supplier No. 70248
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Lightweight wrist support with open center is ideal for repetitive strain and overuse discomfort in office, retail and light industrial applications.

Specifications

Color Black
Size 2X-Large
For Use With (Application) Food Processing, Picking/Packing,Assembly/Fabrication, Office/Retail, Light Industrial
Hand Left
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.