Learn More
New Era Pump Systems Inc PROG MICROFLUIDICS SYRING PUMP
Catalog No. NC1395861
Encompass_Preferred
The smoothest microfluidic syringe pump on the market. Advance Per Step: 4.252232 Nanometers. Holds 1 syringe up to 60 mL (140 mL partially filled). Infusion rates from 0.008 nL/hr (0.5 ¿L syringe) to 2545 ¿L/min (140 mL syringe). Easy-to-use keypad interface. One-click program upload from "Pumping Program Generator" spreadsheet. Space Saving Chassis: Foot print size of only 5 3/4" x 8 3/4". Won't take up unnecessary space on your laboratory or production bench. Dispensing accuracy of +/- 0.5% and reproducibility of +/- 0.2%.
Catalog No. NC1395861 Supplier New Era Pump Systems Inc Supplier No. NE1002X
The smoothest microfluidic syringe pump on the market. Advance Per Step: 4.252232 Nanometers. Holds 1 syringe up to 60 mL (140 mL partially filled). Infusion rates from 0.008 nL/hr (0.5 ¿L syringe) to 2545 ¿L/min (140 mL syringe). Easy-to-use keypad interface. One-click program upload from "Pumping Program Generator" spreadsheet. Space Saving Chassis: Foot print size of only 5 3/4" x 8 3/4". Won't take up unnecessary space on your laboratory or production bench. Dispensing accuracy of +/- 0.5% and reproducibility of +/- 0.2%.