Learn More
Cayman Chemical Progesterone 3-biotin, Purity: ≥98%, 10 mg
SDP
Catalog No. 502893526
Encompass_Preferred
An affinity probe that allows progesterone to be detected or immobilized using the biotin ligand
Catalog No. 50-289-3526 Supplier Cayman Chemical Supplier No. 900064410 MG
An affinity probe that allows progesterone to be detected or immobilized using the biotin ligand