Learn More
Cayman Chemical Progesteron 3-bIotIn 500UG
SDP
Catalog No. 502894499
Encompass_Preferred
An affinity probe that allows progesterone to be detected or immobilized using the biotin ligand
Catalog No. 50-289-4499 Supplier Cayman Chemical Supplier No. 9000644500 UG
An affinity probe that allows progesterone to be detected or immobilized using the biotin ligand