Learn More
Cayman Chemical Progesteron 3-PEG11-bIotIn 1mg
SDP
Catalog No. 502893978
Encompass_Preferred
An affinity probe with a hydrophilic spacer that allows progesterone to be detected or immobilized using the biotin ligand
Catalog No. 50-289-3978 Supplier Cayman Chemical Supplier No. 90006451 MG
An affinity probe with a hydrophilic spacer that allows progesterone to be detected or immobilized using the biotin ligand