Learn More
Cayman Chemical Progesterone 3-PEG11-biotin, Purity: ≥98%, 5 mg
SDP
Catalog No. 502893318
Encompass_Preferred
An affinity probe with a hydrophilic spacer that allows progesterone to be detected or immobilized using the biotin ligand
Catalog No. 50-289-3318 Supplier Cayman Chemical Supplier No. 90006455 MG
An affinity probe with a hydrophilic spacer that allows progesterone to be detected or immobilized using the biotin ligand