Learn More
Cayman Chemical Progesteron 3PEG11bIotIn 10mg
SDP
Catalog No. 502893806
Encompass_Preferred
An affinity probe with a hydrophilic spacer that allows progesterone to be detected or immobilized using the biotin ligand
Catalog No. 50-289-3806 Supplier Cayman Chemical Supplier No. 900064510 MG
An affinity probe with a hydrophilic spacer that allows progesterone to be detected or immobilized using the biotin ligand