Learn More
Medchemexpress LLC Progesterone-13C2 | 82938-07-6 | 316.45 | 1 MG
SDP
Catalog No. 5000389807
Encompass_Preferred
Progesterone-13C2 is the 13C-labeled Progesterone, a steroid hormone that regulates the menstrual cycle and is crucial for pregnancy. It can be used as a tracer or as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
Catalog No. 50-003-89807 Supplier Medchemexpress LLC Supplier No. HYN0437S31MG
Progesterone-13C2 is the 13C-labeled Progesterone, a steroid hormone that regulates the menstrual cycle and is crucial for pregnancy. It can be used as a tracer or as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.