Learn More
Apexbio Technology LLC Progesterone 57-83-0 1g
SDP
Catalog No. 5000095247
Encompass_Preferred
Progesterone (57-83-0) is a steroid hormone that modulates reproductive processes by specifically interacting with its nuclear receptors It is designed to regulate embryo implantation and endometrial physiology thereby influencing estrogenic signaling and target gene expression in reproductive tissues Progesterone exerts its biological activity primarily through attenuation of estrogenic signaling affecting genes such as Egr1 In in vitro studies progesterone demonstrates the ability to restrict cell proliferation pathways in melanoma models including B16F10 mouse cells and BLM human cell lines Additionally progesterone regulates mammary carcinoma progression via RANKL-dependent activation of signaling components such as GLI-1 Based on these pharmacological properties progesterone holds research potential in studies of hormonal regulation reproductive biology cancer progression and neuroprotection
Catalog No. 50-000-95247 Supplier Apexbio Technology LLC Supplier No. A85091000
Progesterone (57-83-0) is a steroid hormone that modulates reproductive processes by specifically interacting with its nuclear receptors It is designed to regulate embryo implantation and endometrial physiology thereby influencing estrogenic signaling and target gene expression in reproductive tissues Progesterone exerts its biological activity primarily through attenuation of estrogenic signaling affecting genes such as Egr1 In in vitro studies progesterone demonstrates the ability to restrict cell proliferation pathways in melanoma models including B16F10 mouse cells and BLM human cell lines Additionally progesterone regulates mammary carcinoma progression via RANKL-dependent activation of signaling components such as GLI-1 Based on these pharmacological properties progesterone holds research potential in studies of hormonal regulation reproductive biology cancer progression and neuroprotection