Learn More
Cayman Chemical Progesterone 5g
SDP
Catalog No. 502972946
Encompass_Preferred
The biosynthetic precursor of all other steroid hormones; activates the human progesterone receptor with an EC50 value of 0.5 nM
Catalog No. 50-297-2946 Supplier Cayman Chemical Supplier No. 158765 G
The biosynthetic precursor of all other steroid hormones; activates the human progesterone receptor with an EC50 value of 0.5 nM