Learn More
Medchemexpress LLC Progesterone carboxylic acid (Etienic acid) | 302-97-6 | 316.43 | 100 MG
SDP
Catalog No. 5000364709
Encompass_Preferred
Progesterone carboxylic acid (also known as Etienic acid or Testosterone-17β-carboxylic acid) is an analogue of androstenedione. This compound has the ability to conjugate with 20-mer peptide nucleic acid (PNA), resulting in a conjugate that exhibits superior binding capacity to complementary DNA. It is actively researched for its potential applications in treating cancers and type 2 diabetes (T2D).
  • Androstenedione analogue.
  • Conjugates with PNA for enhanced DNA binding.
  • Used for cancers and type 2 diabetes research.
Catalog No. 50-003-64709 Supplier Medchemexpress LLC Supplier No. 100MGHY108409
Progesterone carboxylic acid (also known as Etienic acid or Testosterone-17β-carboxylic acid) is an analogue of androstenedione. This compound has the ability to conjugate with 20-mer peptide nucleic acid (PNA), resulting in a conjugate that exhibits superior binding capacity to complementary DNA. It is actively researched for its potential applications in treating cancers and type 2 diabetes (T2D).
  • Androstenedione analogue.
  • Conjugates with PNA for enhanced DNA binding.
  • Used for cancers and type 2 diabetes research.