Learn More
Medchemexpress LLC Progesterone-d9 | 15775-74-3 | 323.52 | 1 MG
SDP
Catalog No. 5000390165
Encompass_Preferred
Progesterone-d9 is a deuterium-labeled version of Progesterone, a steroid hormone vital for regulating the menstrual cycle and pregnancy. Intended for research use only, it can be used as a tracer or an internal standard in quantitative analysis. Deuteration, the incorporation of stable heavy isotopes, can influence the pharmacokinetic and metabolic profiles of drugs.
  • Can be used as a tracer in research applications
  • Functions as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
  • Deuterium labeling can impact pharmacokinetic and metabolic profiles of drugs
  • For research use only, not for patient use
Catalog No. 50-003-90165 Supplier Medchemexpress LLC Supplier No. HYN0437S1MG
Progesterone-d9 is a deuterium-labeled version of Progesterone, a steroid hormone vital for regulating the menstrual cycle and pregnancy. Intended for research use only, it can be used as a tracer or an internal standard in quantitative analysis. Deuteration, the incorporation of stable heavy isotopes, can influence the pharmacokinetic and metabolic profiles of drugs.
  • Can be used as a tracer in research applications
  • Functions as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
  • Deuterium labeling can impact pharmacokinetic and metabolic profiles of drugs
  • For research use only, not for patient use