Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MP Biomedical™ Progesterone Double Antibody RIA Kit

Catalog No. MP07170105 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
100 Tubes
500 Tubes
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MP07170105 500 Tubes
MP07170102 100 Tubes
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MP07170105 Supplier MP Biomedical™ Supplier No. 07170105
Add to Cart
Add to Cart

Quantitative Liquid Phase Radioimmunoassay

Progesterone Double Antibody RIA Kit is intended for the quantitative determination of progesterone in human serum or plasma.
TRUSTED_SUSTAINABILITY

Specifications

Description Intended for the quantitative determination of progesterone in human serum or plasma.
Product Type Kit and Assay
Sample Type Plasma, Serum
Target Species Human
For Use With (Application) Endocrinology
Quantity 500 Tubes
Regulatory Status FDA Cleared (K800126)
Sample Volume 100 μL
Storage Requirements 2°C to 8°C
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.