Learn More
Drg International Inc DRG Progesterone ELISA, 96 Well Kit
Supplier:  Drg International Inc EIA1561
Encompass_Preferred
An enzyme immunoassay for the quantitative determination of progesterone in serum and plasma. This steroid hormone is a female sex hormone which, in conjunction with estrogens, regulates the accessory organs during the menstrual cycle and it is particularly important in preparing the endometrium for the implantation of the blastocyte and in maintaining pregnancy. In non-pregnant women progesterone is mainly secreted by the corpus luteum whereas in pregnancy the placenta becomes the major source. The DRG Progesterone ELISA Kit is a solid phase enzyme-linked immunosorbent assay (ELISA), based on the principle of competitive binding. The microtiter wells are coated with a polyclonal antibody directed towards an antigenic site on the Progesterone molecule.
Catalog No. 50-187-1234