Learn More
Eagle Biosciences Inc Progesterone ELISA
SDP
Supplier:  Eagle Biosciences Inc PRG31K01
Encompass
The Eagle Biosciences Progesterone ELISA Assay Kit (enzyme-linked immunoassay kit) is intended for the direct quantitative determination of progesterone in human serum. The Eagle Biosciences Progesterone ELISA Assay Kit is for research use only and not to be used in diagnostic procedures.
Catalog No. 50-187-8189