Learn More
Enzo Life Sciences Progesterone ELISA kit (5x96wells)
SDP
Supplier:  Enzo Life Sciences ADI901011
Encompass_Preferred
The Progesterone EIA kit is a colorimetric competitive enzyme immunoassay kit with results in < 3 hours. Absorbance is read at 405 nm. This non-radioactive enzyme immunoassay is safe and reliable with ready-to-use liquid color-coded reagents to reduce errors. Sensitivity: 8.57 pg/ml (range 15.62 - 500 pg/ml). Assay Time: <3 hours. Applications: ELISA, Colorimetric detection. Species Reactivity: Species independent. Contents: GxM Microtiter plate, Conjugate, Antibody, Assay buffer concentrate, Wash buffer concentrate, Standard, pNpp Substrate, Stop solution, Steroid displacement reagent
Catalog No. 50-658-203