Learn More
Arbor Assays Progesterone Metabolites ELISA Kit (1 Plate)
SDP
Supplier:  Arbor Assays K068H1
Encompass_Preferred
The DetectX Progesterone Metabolite ELISA Kits quantitatively measure progesterone metabolites present in extracted dried fecal samples and urine.
Catalog No. 50-227-8506