Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Progesterone Metabolites ELISA Kit

Catalog No. EIAPROGM
Encompass_Preferred
Change view
Click to view available options
Quantity:
96 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
EIAPROGM 96 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. EIAPROGM Supplier Invitrogen™ Supplier No. EIAPROGM
Add to Cart
Add to Cart

ELISA

The Progesterone Metabolites Competitive ELISA quantitates progesterone metabolites in serum, plasma, urine, saliva, dried fecal extracts or cell culture medium. Principle of the method: The Progesterone Metabolites Competitive ELISA research-use-only kit is designed to quantitatively measure progesterone metabolites independent of species. A progesterone metabolites standard is provided to generate a standard curve for the assay and all samples are read off a user-generated standard curve. Standards or diluted samples are pipetted into a clear microtiter plate coated with anti-rabbit IgG antibodies. Anti-rabbit progesterone metabolites antibodies and progesterone metabolites-peroxidase conjugate is added to the wells. The progesterone metabolites-peroxidase conjugate and any progesterone metabolites in the sample will compete to bind to the anti-rabbit progesterone metabolites antibodies. After incubation, the plate is washed and substrate is added. The substrate reacts with the bound progesterone metabolites-peroxidase conjugate. After a 30 min incubation, the reaction is stopped and the intensity of the generated color is detected in a microtiter plate reader at 450 nm. The intensity of the generated color corresponds inversely to the amount of progesterone metabolites present in the sample. Rigorous validation: Each manufactured lot of this ELISA kit is quality tested for criteria such as sensitivity, specificity, precision, and lot-to-lot consistency. See manual for m...

Progesterone, C21H30O2, also known as P4 (pregn-4-ene-3,20-dione) is a C-21 steroid hormone involved in the female menstrual cycle, gestation and embryogenesis of humans and other species. Progesterone belongs to a class of hormones called progestogens and is the major naturally occurring human progestogen. Progesterone is an essential regulator of human female reproductive function in the uterus, ovary, mammary gland and brain, and plays an important role in non-reproductive tissues such as the cardiovascular system, bone and the central nervous system.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number C21H30O2; Progesterone Metabolites; progestogen metabolites
Assay Range 102.4-10000 pg/mL
Assay Sensitivity 44.8 pg/mL
Conjugate HRP
Immunoassay Kit Format Sandwich ELISA Kit
Product Type ELISA
Sample Type Fecal Extract, Plasma, Serum, Supernatant, Urine
Target Species Human
For Use With (Equipment) Absorbance Microplate Reader
Gene Alias C21H30O2; Progesterone Metabolites; progestogen metabolitesC21H30O2; Progesterone Metabolites; progestogen metabolites;
Interassay CV 7%
Intraassay CV 7%
Kit Contents Anti-IgG coated 96 well plate, Standard, Assay Buffer Concentrate, Capture Antibody, HRP-conjugated Antigen, Wash Buffer Concentrate, TMB Substrate, Stop Solution, Plate Sealer, Standard, Assay Buffer Concentrate, Capture Antibody, HRP-conjugated Antigen, Wash Buffer Concentrate, TMB Substrate, Stop Solution, Plate Sealer
Label or Dye HRP
Quantity 96 Tests
Regulatory Status RUO
Sample Volume Fecal Extract 50 μL, Plasma 50 μL, Serum 50 μL, Supernatant 50 μL, Urine 12.5 μL
Shipping Condition Wet or Dry Ice
Storage Requirements 2-8°C
Target Progesterone Metabolites
Test Time 45 min
Total Assay Time 1 hr 30 min
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.