Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Progesterone, Micronized Powder, USP, 97-103%, Spectrum™ Chemical
SDP

Catalog No. 18605845 Shop All Spectrum Chemical Manufacturing Corporation Products
Change view
Click to view available options
Quantity:
100 g
500 g
1 kg
2.5 kg
Packaging:
Amber Glass Bottle
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-605-845 500 g Amber Glass Bottle
18-605-848 1 kg Amber Glass Bottle
18-605-846 2.5 kg Amber Glass Bottle
18-605-844 100 g Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 18-605-845 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. P1834500GM
Add to Cart
Add to Cart

P1834, 57-83-0, C21H30O2

Spectrum™ Chemical Progesterone, Micronized Powder, USP is used in hormone replacement therapy and for treating menopausal symptoms. It is manufactured from plants (Yams). All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 57-83-0
Molecular Formula C21H30O2
Molecular Weight (g/mol) 314.47
InChI Key RJKFOVLPORLFTN-LEKSSAKUSA-N
IUPAC Name (1S,3aS,3bS,9aR,9bS,11aS)-1-acetyl-9a,11a-dimethyl-1H,2H,3H,3aH,3bH,4H,5H,7H,8H,9H,9aH,9bH,10H,11H,11aH-cyclopenta[a]phenanthren-7-one
SMILES CC(=O)[C@H]1CC[C@H]2[C@@H]3CCC4=CC(=O)CC[C@]4(C)[C@H]3CC[C@]12C

Specifications

CAS Max % 1
Loss on Drying 0.005
Melting Range 126-131°C
Packaging Amber Glass Bottle
Quantity 500 g
Specific Rotation +175° to +183°
Percent Purity 97 to 103%
Grade USP
Chemical Name or Material Progesterone
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.