Learn More
Selleck Chemical LLC Progesterone 25mg 57-83-0
SDP
Supplier:  Selleck Chemical LLC S170525MG
Encompass_Preferred
Progesterone is an endogenous steroid hormone involved in the menstrual cycle, pregnancy, and embryogenesis of humans and other species. A potent agonist of the nuclear progesterone receptor (nPR) with Kd of 1 nM; An agonist of the membrane progesterone receptors(mPRs); An antagonist of the ?1 receptor. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-572-0