Learn More
Cayman Chemical Progesterone Receptor Phosph
SDP
Supplier:  Cayman Chemical 100097631 EA
Encompass_Preferred
Antigen: phosphopeptide corresponding to amino acid residues surrounding the phospho-Ser294 of human progesterone receptor · Host: mouse · Cross-reactivity: (+) human progesterone receptor; expected to react with mouse, rat, non-human primates progesterone receptor based on 100% homology with the amino acid sequence used as the antigen · Application: WB and IHC (frozen sections)
Catalog No. 50-288-7030