Learn More
Cayman Chemical Progesterone Receptor Phosph
SDP
Catalog No. 502886599
Encompass_Preferred
Immunogen: Phosphopeptide from amino acid residues surrounding the phospho-Ser294 of human progesterone receptor • Host: Mouse • Species Reactivity: (+) Human • Applications: IHC, WB
Catalog No. 50-288-6599 Supplier Cayman Chemical Supplier No. 100097621 EA
Immunogen: Phosphopeptide from amino acid residues surrounding the phospho-Ser294 of human progesterone receptor • Host: Mouse • Species Reactivity: (+) Human • Applications: IHC, WB