Learn More
Bioss Progesterone Receptor - Ser190
SDP
Catalog No. 5000065139
Encompass_Preferred
Progesterone Receptor (Ser190) Primary Antibody Unconjugated Host Species Mouse Target Species Human Clonality Monoclonal Applications WB IHC 1154Concentration Lot Dependent Size 100 ul Storage Buffer 10 mM HEPES (pH 7 5) 150 mM NaCl 100 g per ml BSA and 50% glycerol
Catalog No. 50-000-65139 Supplier Bioss Supplier No. BSM70231M
Progesterone Receptor (Ser190) Primary Antibody Unconjugated Host Species Mouse Target Species Human Clonality Monoclonal Applications WB IHC 1154Concentration Lot Dependent Size 100 ul Storage Buffer 10 mM HEPES (pH 7 5) 150 mM NaCl 100 g per ml BSA and 50% glycerol