Learn More
Bioss Progesterone Receptor - Ser294
SDP
Catalog No. 5000064731
Encompass_Preferred
Progesterone Receptor (Ser294) Primary Antibody Unconjugated Host Species Mouse Target Species Human Clonality Monoclonal Applications WB IHC 608Concentration Lot Dependent Size 100 ul Storage Buffer 10 mM HEPES (pH 7 5) 150 mM NaCl 100 g per ml BSA and 50% glycerol
Catalog No. 50-000-64731 Supplier Bioss Supplier No. BSM70232M
Progesterone Receptor (Ser294) Primary Antibody Unconjugated Host Species Mouse Target Species Human Clonality Monoclonal Applications WB IHC 608Concentration Lot Dependent Size 100 ul Storage Buffer 10 mM HEPES (pH 7 5) 150 mM NaCl 100 g per ml BSA and 50% glycerol