Learn More
Description
Progesterone USP is a white or creamy white, crystalline powder. It is stable in air. Check for meaning of milled, why used for this version.
Specifications
Specifications
| Chemical Name or Material | Progesterone USP (Milled) |
| Packaging | Jar, 2.8L, White, HDPE, Round, 110-400, TE Pano Neck |
| CAS | 57-83-0 |
| Type | Drug Standard |
| Molecular Weight (g/mol) | 314.46 |
| Quantity | 1 kg |
| Percent Purity | 97 to 103 % |
| Grade | USP |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
