Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medisca Progesterone USP (Milled)
SDP

Catalog No. 14185416 Shop All Medisca Products
Change view
Click to view available options
Quantity:
100 g
1 kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
14-185-416 100 g
14-185-191 1 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 14-185-416 Supplier Medisca Supplier No. 031005
Only null left
Add to Cart
Add to Cart

Progesterone USP is a white or creamy white, crystalline powder. It is stable in air. Check for meaning of milled, why used for this version.

Progesterone USP is a white or creamy white, crystalline powder. It is stable in air. Check for meaning of milled, why used for this version.

Specifications

Chemical Name or Material Progesterone USP (Milled)
Packaging Jar, 300mL, White, HDPE, Round, 63-400, TE Pano Neck
CAS 57-83-0
Type Drug Standard
Molecular Weight (g/mol) 314.46
Quantity 100 g
Percent Purity 97 to 103 %
Grade USP
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.