Learn More
Cayman Chemical Progesterone-3-biotinyl Photoaffinity Probe, Purity: ≥95% (mixture of isomers), 5 mg
SDP
Catalog No. 502893548
Encompass_Preferred
A photoaffinity probe that consists of active progesterone, a diazirine-UV (312 nm)-sensitive moiety, and a biotin tag; can be used to detect and observe the interaction of progesterone with target proteins
Catalog No. 50-289-3548 Supplier Cayman Chemical Supplier No. 90008915 MG
A photoaffinity probe that consists of active progesterone, a diazirine-UV (312 nm)-sensitive moiety, and a biotin tag; can be used to detect and observe the interaction of progesterone with target proteins