Learn More
TARGETMOL CHEMICALS INC Proglumide 200MG
SDP
Catalog No. 502880522
Encompass_Preferred
Also available in 1 mL, 25 mg, 50 mg, 100 mg, 500 mg and bulk. Please contact Fisher for quotes. Proglumide (Binoside) is a cholecystokinin antagonist, which blocks both the CCKA and CCKB subtypes. Proglumide is a drug that exerts an inhibitory effect on gastric secretion and reduces gastrointestinal motility. It is used clinically in the drug therapy of gastrointestinal ulcers. Purity 99.44%
Catalog No. 50-288-0522 Supplier TARGETMOL CHEMICALS INC Supplier No. T1052200MG
Also available in 1 mL, 25 mg, 50 mg, 100 mg, 500 mg and bulk. Please contact Fisher for quotes. Proglumide (Binoside) is a cholecystokinin antagonist, which blocks both the CCKA and CCKB subtypes. Proglumide is a drug that exerts an inhibitory effect on gastric secretion and reduces gastrointestinal motility. It is used clinically in the drug therapy of gastrointestinal ulcers. Purity 99.44%