Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Proglumide sodium | 99247-33-3 | MFCD00069301 | 99.6% | 356.39 g·mol⁻¹ | C18H25N2NaO4 | 1 ML
SDP

Catalog No. 5000245051
Encompass_Preferred

Proglumide sodium is the sodium salt of proglumide, a nonpeptide cholecystokinin (CCK) A/B receptor antagonist used as a research reagent. Supplied as an aqueous solution at 10 mM concentration in a 1 mL vial, it is intended for in vitro research and receptor pharmacology studies.

  • Ready-to-use 10 mM aqueous solution.
  • Supplied in a 1 mL vial for small-volume assays.
  • Molecular weight 356.39 g·mol⁻¹.
  • Molecular formula C18H25N2NaO4.
  • Reported purity 99.6% suitable for research applications.
  • Useful for CCK-A/B receptor modulation and related pharmacology studies.

Catalog No. 50-002-45051 Supplier Medchemexpress LLC Supplier No. HY10335410MM 1ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Proglumide sodium is the sodium salt of proglumide, a nonpeptide cholecystokinin (CCK) A/B receptor antagonist used as a research reagent. Supplied as an aqueous solution at 10 mM concentration in a 1 mL vial, it is intended for in vitro research and receptor pharmacology studies.

  • Ready-to-use 10 mM aqueous solution.
  • Supplied in a 1 mL vial for small-volume assays.
  • Molecular weight 356.39 g·mol⁻¹.
  • Molecular formula C18H25N2NaO4.
  • Reported purity 99.6% suitable for research applications.
  • Useful for CCK-A/B receptor modulation and related pharmacology studies.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.