Learn More
Medchemexpress LLC Progoitrin | 585-95-5 | 98.7% | 389.40 g/mol | C11H19NO10S2 | 10 MG
SDP
Catalog No. 5000295776
Encompass_Preferred
Progoitrin is a glucosinolate compound found in certain crops and reported to have antithyroid activity. Provided as a white to off-white solid for research use, it has CAS 585-95-5 and molecular formula C11H19NO10S2. Store sealed at 4°C, protected from moisture and light, and consult the lot-specific COA for exact purity and handling information.
  • High reported purity suitable for analytical and biochemical studies.
  • Solid form with white to off-white appearance for convenient handling.
  • Stability in solution: -80°C for up to 6 months, -20°C for up to 1 month (sealed).
  • Available in small milligram quantities for laboratory experiments.
Catalog No. 50-002-95776 Supplier Medchemexpress LLC Supplier No. HYN1034810MG
Progoitrin is a glucosinolate compound found in certain crops and reported to have antithyroid activity. Provided as a white to off-white solid for research use, it has CAS 585-95-5 and molecular formula C11H19NO10S2. Store sealed at 4°C, protected from moisture and light, and consult the lot-specific COA for exact purity and handling information.
  • High reported purity suitable for analytical and biochemical studies.
  • Solid form with white to off-white appearance for convenient handling.
  • Stability in solution: -80°C for up to 6 months, -20°C for up to 1 month (sealed).
  • Available in small milligram quantities for laboratory experiments.