Learn More
Cole-Parmer Cole-Parmer® OVV-400 Series Programmable Vacuum Ovens
Catalog No. NC3791906 Shop All Cole-Parmer Products
Encompass_Preferred
Easily manage up to eight programs with multiple steps each. -Stainless steel inner chamber resists corrosion. -Rounded inner chamber corners offer easy cleaning. -Built-in alarm alerts when oven shuts down. -Able to heat samples up to 392°F (200°C)
Catalog No. NC3791906 Supplier Cole-Parmer Supplier No. 5241130
Easily manage up to eight programs with multiple steps each. -Stainless steel inner chamber resists corrosion. -Rounded inner chamber corners offer easy cleaning. -Built-in alarm alerts when oven shuts down. -Able to heat samples up to 392°F (200°C)