Learn More
Biomatik Corporation Programmed Cell Death Protein 1 Ligand 1 (Pdl1) Monoclonal Antibody
Supplier:  Biomatik Corporation CAU29803
Encompass_Preferred
Programmed Cell Death Protein 1 Ligand 1 (Pdl1) Monoclonal Antibody is a Human (Homo sapiens) Monoclonal Antibody to Programmed Cell Death Protein 1 Ligand 1 (PDL1). The Programmed Cell Death Protein 1 Ligand 1 (Pdl1) Monoclonal Antibody has been validated for the following applications: WB, IHC, ICC, IP. Host: Mouse. Species: Human (Homo sapiens). Immunogen: Pro24~Pro234. Applications: WB, IHC, ICC, IP. Target Synonyms: Programmed Cell Death Protein 1 Ligand 1 (PDL1). Accession Number: Q9NZQ7. Gene ID: 29126. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-7199