Learn More
Crystal Chem Inc Progranulin ELISA Kit, 96-wells
SDP
Supplier:  Crystal Chem Inc 80590
Encompass
The Progranulin ELISA is used to quantify levels of human progranulin in plasma, serum, and cell culture using only a 10 µL sample volume. Highlights: Kit uses sample size of only 10 µL. Dynamic range of 0.075 - 25 ng/mL. High sensitivity of 0.018 ng/mL. Crystal Chem is a worldwide leading supplier of ELISA kits for human markers. Our kits are among the highest quality found in the market, and are used by leading research universities, pharma companies, and government institutions around the world.
Catalog No. 50-194-7756