Learn More
AdipoGen Progranulin h untagged
SDP
Supplier:  AdipoGen AG40A0188YC010
Encompass_Preferred
Protein. Signal peptide and human progranulin aa 1-593 is untagged. Reflects the native sequence with no additional aa. Source HEK 293 cells. Endotoxin content <0.01EU/µg purified protein LAL test Lonza. Lyophilized. Contains PBS. Purity >98% SDS-PAGE. Progranulin PGRN is a widely expressed pluripotent growth factor which plays a role in processes such as development, wound repair and inflammation by activating signaling cascades that control cell cycle progression and cell motility. Its function in the central nervous system is of interest, as mutations in the PGRN gene were found in cases of frontotemporal degeneration FTLD. In addition, PGRN has also been linked to tumorigenesis. Progranulin is a biomarker for FTLD, other types of Alzheimer‘s Disease AD and potentially for MCI Mild Cognitive Impairment. Additionally, PGRN is described as a new ligand of TNF receptors and a potential therapeutic against inflammatory disease like arthritis.
Catalog No. 50-256-2955