Learn More
AdipoGen Progranulin (mouse) ELISA Kit,ELISA Assay
SDP
Supplier:  AdipoGen AG45A0019YEKKI01
Encompass_Preferred
Progranulin (mouse) ELISA Kit, AdipoGen Life Sciences. ELISA Assay. Detects mouse progranulin. Does not cross-react with human progranulin. Colorimetric assay. Sample Types: Cell Culture Supernatant, Serum. Range: 0.125 to 8ng/ml. Sensitivity: 60pg/ml. Progranulin (PGRN) is a widely expressed pluripotent growth factor which plays a role in processes such as development, wound repair and inflammation by activating signaling cascades that control cell cycle progression and cell motility. Its function in the central nervous system is of interest, as mutations in the PGRN gene were found in cases of frontotemporal degeneration (FTLD). In addition, PGRN has also been linked to tumorigenesis. New cutting-edge publications have shown that progranulin might be a biomarker not only for FTLD, but also for other types of Alzheimer¿s disease (AD). This new data even supposes the potential of progranulin to be a biomarker for MCI, which would be a hallmark in AD research.
Catalog No. 50-101-5787