Learn More
AdipoGen Progranulin mouse pAb
SDP
Supplier:  AdipoGen AG25A0093C100
Encompass_Preferred
Polyclonal Antibody. Recognizes mouse progranulin. Detects a band of ~90kDa by Western blot. Weakly cross-reacts with human progranulin. Source Rat. Applications ELISA, WB. Liquid. 0.2µm-filtered solution in PBS, pH 7.4. Contains no preservatives. Progranulin PGRN is a widely expressed pluripotent growth factor which plays a role in processes such as development, wound repair and inflammation by activating signaling cascades that control cell cycle progression and cell motility. Its function in the central nervous system is of interest, as mutations in the PGRN gene were found in cases of frontotemporal degeneration FTLD. In addition, PGRN has also been linked to tumorigenesis. Progranulin is a biomarker for FTLD, other types of Alzheimer‘s Disease AD and potentially for MCI Mild Cognitive Impairment. Additionally, PGRN is described as a new ligand of TNF receptors and a potential therapeutic against inflammatory disease like arthritis.
Catalog No. 50-256-3227