Learn More
Medchemexpress LLC Progranulin/PGRN Mo 100ug | 100ug
SDP
Catalog No. 5000193907
Encompass_Preferred
Progranulin (PGRN) proteins regulate lysosomal function affecting protein transport enzymatic activity and acidification It degrades mature cathepsin D via cathepsin B Progranulin/PGRN Protein Mouse (572a a HEK293 His) is the recombinant mouse-derived Progranulin/PGRN protein expressed by HEK293 with C-His labeled tag
Catalog No. 50-001-93907 Supplier Medchemexpress LLC Supplier No. HYP78302100UG
Progranulin (PGRN) proteins regulate lysosomal function affecting protein transport enzymatic activity and acidification It degrades mature cathepsin D via cathepsin B Progranulin/PGRN Protein Mouse (572a a HEK293 His) is the recombinant mouse-derived Progranulin/PGRN protein expressed by HEK293 with C-His labeled tag