Learn More
Medchemexpress LLC Progranulin/PGRN Mo 20ug | 20ug
SDP
Catalog No. 5000193711
Encompass_Preferred
Progranulin (PGRN) proteins regulate lysosomal function affecting protein transport enzymatic activity and acidification It degrades mature cathepsin D via cathepsin B Progranulin/PGRN Protein Mouse (572a a HEK293 His) is the recombinant mouse-derived Progranulin/PGRN protein expressed by HEK293 with C-His labeled tag
Catalog No. 50-001-93711 Supplier Medchemexpress LLC Supplier No. HYP7830220UG
Progranulin (PGRN) proteins regulate lysosomal function affecting protein transport enzymatic activity and acidification It degrades mature cathepsin D via cathepsin B Progranulin/PGRN Protein Mouse (572a a HEK293 His) is the recombinant mouse-derived Progranulin/PGRN protein expressed by HEK293 with C-His labeled tag