Learn More
AdipoGen Progranulin rat rec
SDP
Catalog No. 502562946
Encompass_Preferred
Protein. Signal peptide and rat progranulin aa 1-602 are fused at the C-terminus to a FLAG -tag. Source HEK 293 cells. Endotoxin content <0.1EU/µg purified protein LAL test Lonza. Lyophilized from 0.2µm-filtered solution in PBS. Purity >95% SDS-PAGE. Progranulin PGRN is a widely expressed pluripotent growth factor which plays a role in processes such as development, wound repair and inflammation by activating signaling cascades that control cell cycle progression and cell motility. Its function in the central nervous system is of interest, as mutations in the PGRN gene were found in cases of frontotemporal degeneration FTLD. In addition, PGRN has also been linked to tumorigenesis. Progranulin is a biomarker for FTLD, other types of Alzheimer‘s Disease AD and potentially for MCI Mild Cognitive Impairment. Additionally, PGRN is described as a new ligand of TNF receptors and a potential therapeutic against inflammatory disease like arthritis.
Catalog No. 50-256-2946 Supplier AdipoGen Supplier No. AG40A0194C010
Protein. Signal peptide and rat progranulin aa 1-602 are fused at the C-terminus to a FLAG -tag. Source HEK 293 cells. Endotoxin content <0.1EU/µg purified protein LAL test Lonza. Lyophilized from 0.2µm-filtered solution in PBS. Purity >95% SDS-PAGE. Progranulin PGRN is a widely expressed pluripotent growth factor which plays a role in processes such as development, wound repair and inflammation by activating signaling cascades that control cell cycle progression and cell motility. Its function in the central nervous system is of interest, as mutations in the PGRN gene were found in cases of frontotemporal degeneration FTLD. In addition, PGRN has also been linked to tumorigenesis. Progranulin is a biomarker for FTLD, other types of Alzheimer‘s Disease AD and potentially for MCI Mild Cognitive Impairment. Additionally, PGRN is described as a new ligand of TNF receptors and a potential therapeutic against inflammatory disease like arthritis.