Learn More
Selleck Chemical LLC Proguanil
SDP
Catalog No. 5000003474
Encompass_Preferred
Proguanil (chlorguanide chloroguanide) is a prophylactic antimalarial drug It inhibits the dihydrofolate reductase of plasmodia and thereby blocks the biosynthesis of purines and pyrimidines which are essential for DNA synthesis and cell multiplication
Catalog No. 50-000-03474 Supplier Selleck Chemical LLC Supplier No. S592725MG
Proguanil (chlorguanide chloroguanide) is a prophylactic antimalarial drug It inhibits the dihydrofolate reductase of plasmodia and thereby blocks the biosynthesis of purines and pyrimidines which are essential for DNA synthesis and cell multiplication