Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Proguanil-d6 | 99.31% | 500 UG
SDP

Catalog No. 5000409227
Encompass_Preferred

Proguanil-d6 is a deuterium-labeled prophylactic antimalarial agent, derived from Proguanil.

  • Functions as a tracer
  • Serves as an internal standard for quantitative analysis via NMR, GC-MS, or LC-MS
  • Targets Plasmodium
  • Solid appearance, ranging from white to off-white
  • Molecular formula: C11H10D6ClN5
  • Molecular weight: 259.77

Catalog No. 50-004-09227 Supplier Medchemexpress LLC Supplier No. HYB0806S500UG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Proguanil-d6 is a deuterium-labeled prophylactic antimalarial agent, derived from Proguanil.

  • Functions as a tracer
  • Serves as an internal standard for quantitative analysis via NMR, GC-MS, or LC-MS
  • Targets Plasmodium
  • Solid appearance, ranging from white to off-white
  • Molecular formula: C11H10D6ClN5
  • Molecular weight: 259.77

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.