Learn More
Medchemexpress LLC Proguanil-d6 (hydrochloride) | 1 MG
SDP
Catalog No. 5000408037
Encompass_Preferred
Proguanil-d6 (hydrochloride) is a deuterium-labeled form of Proguanil hydrochloride, an antimalarial proagent metabolized to the active metabolite Cycloguanil. It functions as a dihydrofolate reductase (DHFR) inhibitor. This product is a solid, intended for research use only, and suitable for use as a laboratory chemical in the manufacture of substances.
  • Deuterium-labeled compound
  • Functions as an antimalarial proagent
  • Metabolized to the active metabolite cycloguanil
  • Dihydrofolate reductase (DHFR) inhibitor
  • Useful as a laboratory chemical
  • Appears as a solid
  • Intended for research use only
Catalog No. 50-004-08037 Supplier Medchemexpress LLC Supplier No. HYB0806AS11MG
Proguanil-d6 (hydrochloride) is a deuterium-labeled form of Proguanil hydrochloride, an antimalarial proagent metabolized to the active metabolite Cycloguanil. It functions as a dihydrofolate reductase (DHFR) inhibitor. This product is a solid, intended for research use only, and suitable for use as a laboratory chemical in the manufacture of substances.
  • Deuterium-labeled compound
  • Functions as an antimalarial proagent
  • Metabolized to the active metabolite cycloguanil
  • Dihydrofolate reductase (DHFR) inhibitor
  • Useful as a laboratory chemical
  • Appears as a solid
  • Intended for research use only