Learn More
Medchemexpress LLC Proguanil-d6 hydrochloride | 500-92-5 | 99.31% | 5 MG
SDP
Catalog No. 5000409271
Encompass_Preferred
Proguanil-d6 hydrochloride is the deuterium labeled Proguanil hydrochloride, an antimalarial proagent that is metabolized to the active metabolite Cycloguanil. It acts as a dihydrofolate reductase (DHFR) inhibitor. Stable heavy isotopes, such as deuterium, are incorporated into drug molecules to serve as tracers for quantitation during drug development and may impact pharmacokinetic and metabolic profiles.
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
Catalog No. 50-004-09271 Supplier Medchemexpress LLC Supplier No. HYB0806AS15MG
Proguanil-d6 hydrochloride is the deuterium labeled Proguanil hydrochloride, an antimalarial proagent that is metabolized to the active metabolite Cycloguanil. It acts as a dihydrofolate reductase (DHFR) inhibitor. Stable heavy isotopes, such as deuterium, are incorporated into drug molecules to serve as tracers for quantitation during drug development and may impact pharmacokinetic and metabolic profiles.
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS