Learn More
Enterprise Technology Solutions ProGuard Respirator, f/Dust/Mist, Double-shell, 20/BX, White
SDP
Catalog No. 5000828357
Encompass_Preferred
Dust and mist respirator helps you avoid breathing nontoxic particles while working. Double-shell construction protects you from the effects of pollen, dust, grass and more. Molded shape provides a tight-fitting seal to prevent particles from getting through. Custom-fit nosepiece sits comfortably on your face to allow extended use. N-95 rated respirator is OSHA and NIOSH approved TC-21C-271.
Catalog No. 50-008-28357 Supplier Enterprise Technology Solutions Supplier No. PGD7312B
Dust and mist respirator helps you avoid breathing nontoxic particles while working. Double-shell construction protects you from the effects of pollen, dust, grass and more. Molded shape provides a tight-fitting seal to prevent particles from getting through. Custom-fit nosepiece sits comfortably on your face to allow extended use. N-95 rated respirator is OSHA and NIOSH approved TC-21C-271.